Merck
  • Racemization of the aspartic acid residue of amyloid-β peptide by a radical reaction.
Racemization of the aspartic acid residue of amyloid-β peptide by a radical reaction.
Bioscience, biotechnology, and biochemistry (2013-02-09)
Koharu Tambo, Tomomi Yamaguchi, Keiko Kobayashi, Eri Terauchi, Ikuyo Ichi, Shosuke Kojo
ABSTRACT
Human amyloid-β peptide 1-42 (Aβ) was subjected to a radical reaction by using ascorbic acid and CuCl(2). The percentage of D-aspartic acid (D-Asp) after 24 h had increased to 6.69 ± 0.09%, this being comparable with the reported D-Asp concentration of purified core amyloids in Alzheimer's disease patients. This racemization was significantly inhibited by radical scavengers. L-Alanine was also racemized during the same reaction.
Sigma-Aldrich
Copper(II) chloride, 97%
Sigma-Aldrich
Copper(II) chloride, powder, 99%
Sigma-Aldrich
Copper(II) chloride, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Copper(II) chloride, 99.999% trace metals basis
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Copper(II) chloride dihydrate, 99.999%
Sigma-Aldrich
Copper(II) chloride dihydrate, ≥99.95% trace metals basis
Sigma-Aldrich
Copper(II) chloride dihydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Copper(II) chloride dihydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Copper(II) chloride dihydrate, reagent grade