Merck
  • Metabolomic and transcriptomic profiling of human K-ras oncogene transgenic rats with pancreatic ductal adenocarcinomas.
Metabolomic and transcriptomic profiling of human K-ras oncogene transgenic rats with pancreatic ductal adenocarcinomas.
Carcinogenesis (2013-02-09)
Setsuko Yabushita, Katsumi Fukamachi, Hajime Tanaka, Takako Fukuda, Kayo Sumida, Yoshihito Deguchi, Kazuki Mikata, Kazuhiko Nishioka, Satoshi Kawamura, Satoshi Uwagawa, Masumi Suzui, David B Alexander, Hiroyuki Tsuda
ABSTRACT
Pancreatic ductal adenocarcinoma (PDAC) is one of the most debilitating malignancies in humans, and one of the reasons for this is the inability to diagnose this disease early in its development. To search for biomarkers that can be used for early diagnosis of PDAC, we established a rat model of human PDAC in which expression of a human K-ras(G12V) oncogene and induction of PDAC are regulated by the Cre/lox system. In the present study, transgenic rats bearing PDAC and control transgenic rats with normal pancreatic tissues were used for metabolomic analysis of serum and pancreatic tissue by non-targeted and targeted gas chromatography-mass spectrometry and transcriptomic analysis of pancreatic tissue by microarray. Comparison of the metabolic profiles of the serum and pancreatic tissue of PDAC-bearing and control rats identified palmitoleic acid as a metabolite, which was significantly decreased in the serum of PDAC-bearing animals. Transcriptomic analysis indicated that several transcripts involved in anaerobic glycolysis and nucleotide degradation were increased and transcripts involved in the trichloroacetic acid cycle were decreased. Other transcripts that were changed in PDAC-bearing rats were adenosine triphosphate citrate lyase (decreased: fatty acid biosynthesis), fatty acid synthase (increased: fatty acid biosynthesis) and arachidonate 5-lipoxygenase activating protein (increased: arachidonic acid metabolism). Overall, our results suggest that the decreased serum levels of palmitoleic acid in rats with PDAC was likely due to its decrease in pancreatic tissue and that palmitoleic acid should be investigated in human samples to assess its diagnostic significance as a serum biomarker for human PDAC.
Sigma-Aldrich
Palmitoleic acid, ≥98.5% (GC), liquid
Supelco
Palmitoleic acid, analytical standard