Merck
  • Prediction of micelle/water and liposome/water partition coefficients based on molecular dynamics simulations, COSMO-RS, and COSMOmic.
Prediction of micelle/water and liposome/water partition coefficients based on molecular dynamics simulations, COSMO-RS, and COSMOmic.
Langmuir : the ACS journal of surfaces and colloids (2013-02-13)
Thomas Ingram, Sandra Storm, Linda Kloss, Tanja Mehling, Sven Jakobtorweihen, Irina Smirnova
ABSTRACT
Liposomes and micelles find various applications as potential solubilizers in extraction processes or in drug delivery systems. Thermodynamic and transport processes governing the interactions of different kinds of solutes in liposomes or micelles can be analyzed regarding the free energy profiles of the solutes in the system. However, free energy profiles in heterogeneous systems such as micelles are experimentally almost not accessible. Therefore, the development of predictive methods is desirable. Molecular dynamics (MD) simulations reliably simulate the structure and dynamics of lipid membranes and micelles, whereas COSMO-RS accurately reproduces solvation free energies in different solvents. For the first time, free energy profiles in micellar systems, as well as mixed lipid bilayers, are investigated, taking advantage of both methods: MD simulations and COSMO-RS, referred to as COSMOmic (Klamt, A.; Huniar, U.; Spycher, S.; Keldenich, J. COSMOmic: A Mechanistic Approach to the Calculation of Membrane-Water Partition Coefficients and Internal Distributions within Membranes and Micelles. J. Phys. Chem. B 2008, 112, 12148-12157). All-atom molecular dynamics simulations of the system SDS/water and CTAB/water have been applied in order to retrieve representative micelle structures for further analysis with COSMOmic. For the system CTAB/water, different surfactant concentrations were considered, which results in different micelle sizes. Free energy profiles of more than 200 solutes were predicted and validated by means of experimental partition coefficients. To our knowledge, these are the first quantitative predictions of micelle/water partition coefficients, which are based on whole free energy profiles from molecular methods. Further, the partitioning in lipid bilayer systems containing different hydrophobic tail groups (DOPC (1,2-dioleoyl-sn-glycero-3-phosphocholine), SOPC (stearoyl-oleoylphosphatidylcholine), DMPC (1,2-dimyristoyl-sn-glycero-3-phosphocholine), and POPC (1-palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine)) as well as mixed bilayers was calculated. Experimental partition coefficients (log P) were reproduced with a root-mean-square error (RMSE) of 0.62. To determine the influence of cholesterol as an important component of cellular membranes, free energy profiles in the presence of cholesterol were calculated and shown to be in good agreement with experimental data.
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O