Merck
  • Improving the performance of membrane bioreactors by adding a metabolic uncoupler, 4-nitrophenol.
Improving the performance of membrane bioreactors by adding a metabolic uncoupler, 4-nitrophenol.
Applied biochemistry and biotechnology (2013-02-13)
Tang Qiong, Lin Song, Ying Cheng, Jun-Ru Xiong
ABSTRACT
Effects of 4-nitrophenol (4NP) on the overall performance of membrane bioreactors (MBRs) were investigated in two bench-scale submerged MBRs. Positive impacts of 4NP on activated sludge production and membrane fouling were demonstrated over 45 days of stable operational period. After addition of 4-nitrophenol, the sludge production could be reduced effectively, but only a slight reduction in chemical oxygen demand removal was obtained. The effluent NH4(+)-N concentrations were almost the same in two MBRs. The transmembrane pressures (TMPs) and resistance R increased with increasing mixed liquor suspended solid concentration at each MBR. The average daily TMP increase rates in the control MBR reactor remained at about 0.23 kPa day(-1) and dropped to about 0.12 kPa day(-1) in the 4NP-MBR. Compared with the control MBR, a wider dispersion and lower peak of floc size, a lower zeta potential, and a lower extracellular polymeric substance concentration were observed in the 4NP-MBR.
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Supelco
Acetaminophen Related Compound F, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
4-Nitrophenol, PESTANAL®, analytical standard
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Sigma-Aldrich
4-Nitrophenol, spectrophotometric grade