Merck
  • [Modern approaches to the treatment of biliary pathology in young patients with obesity].
[Modern approaches to the treatment of biliary pathology in young patients with obesity].
Eksperimental'naia i klinicheskaia gastroenterologiia = Experimental & clinical gastroenterology (2013-02-14)
G Ia Khismatullina, L V Volevach, G A Khakamova, V V Uliamaeva, M A Bychenkova
ABSTRACT
The therapeutic efficiency of ursosan in 64 overweight patients with chronic noncalculous cholecystitis and dysfunction of biliary tract was evaluated. Including of ursosan in the complex therapy of patients with chronic noncalculous cholecystitis enhances the efficiency of therapy, causes the positive changes in clinical symptoms and indicators of the biliary tract function, diminishes the lithogenic properties of bile, normalize the food behavior.
Product Description
Ursodeoxycholic acid for system suitability, European Pharmacopoeia (EP) Reference Standard
Ursodeoxycholic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ursodeoxycholic acid, ≥99%