Merck
Clinical Inquiry: which treatments help women with reduced libido?
The Journal of family practice (2013-02-14)
Keia Hobbs, Lara Handler
ABSTRACT
Several treatments produce modest, but statistically significant, clinical increases in sexual desire and function in women. The testosterone transdermal patch improves hypoactive sexual desire disorder (HSDD) in postmenopausal women.
Sigma-Aldrich
Bupropion hydrochloride, ≥98% (HPLC), solid
Supelco
Bupropion hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®