Merck
  • Vitexin-2-O-xyloside, raphasatin and (-)-epigallocatechin-3-gallate synergistically affect cell growth and apoptosis of colon cancer cells.
Vitexin-2-O-xyloside, raphasatin and (-)-epigallocatechin-3-gallate synergistically affect cell growth and apoptosis of colon cancer cells.
Food chemistry (2013-02-16)
Alessio Papi, Fulvia Farabegoli, Renato Iori, Marina Orlandi, Gina R De Nicola, Manuela Bagatta, Donato Angelino, Lorenzo Gennari, Paolino Ninfali
ABSTRACT
Cytotoxic effects of the combination of the food components vitexin-2-O-xyloside (X), raphasatin (4-methylsulphanyl-3-butenyl isothiocyanates; G) and (-)-epigallocatechin-3-gallate (E) were investigated in colon (LoVo and CaCo-2) and breast (MDA-MB-231 and MCF-7) cancer cells. Breast cancer cells were more resistant than colon cells to X, G and E inhibition. On the contrary, marked synergistic effects among X, G and E on cell growth were found in both colon cancer cells. Further analysis revealed a G0/G1 arrest of the phase cell progression and apoptosis, linked to modulation of Bax, Bcl2, caspase-9 and poly(ADP-ribose) polymerase as well as Reactive Oxygen Species (ROS) generation in both colon cancer cells, whereas apoptosis and ROS were not significantly detected in normal human lymphocytes. We conclude that the X, G and E mixture might act by mitochondrial pathway activation of apoptosis, possibly elicited by ROS and the mixture may be effective in the chemoprevention of colon cancer.
Product Description
Supelco
Apigenin, analytical standard
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Supelco
(−)-Epigallocatechin gallate, analytical standard
Epigallocatechin gallate, primary reference standard
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea