Merck
  • Novel linear and step-gradient counter-current chromatography for bio-guided isolation and purification of cytotoxic podophyllotoxins from Dysosma versipellis (Hance).
Novel linear and step-gradient counter-current chromatography for bio-guided isolation and purification of cytotoxic podophyllotoxins from Dysosma versipellis (Hance).
Journal of separation science (2013-02-19)
Zhi Yang, Xiaoman Liu, Kuiwu Wang, Xiaoji Cao, Shihua Wu
ABSTRACT
Dysosma versipellis (Hance) is a famous traditional Chinese medicine for the treatment of snakebite, weakness, condyloma accuminata, lymphadenopathy, and tumors for thousands of years. In this work, four podophyllotoxin-like lignans including 4'-demethylpodophyllotoxin (1), α-peltatin (2), podophyllotoxin (3), β-peltatin (4) as major cytotoxic principles of D. versipellis were successfully isolated and purified by several novel linear and step gradient counter-current chromatography methods using the systems of hexane/ethyl acetate/methanol/water (4:6:3:7 and 4:6:4:6, v/v/v/v). Compared with isocratic elution, linear and step-gradient elution can provide better resolution and save more time for the separation of photophyllotoxin and its congeners. Their cytotoxicities were further evaluated and their structures were validated by high-resolution electrospray TOF MS and nuclear magnetic resonance spectra. All components showed potent anticancer activity against human hepatoma cells HepG2.
Sigma-Aldrich
Picropodophyllotoxin, ≥96% (HPLC), powder
Sigma-Aldrich
Podophyllotoxin