Merck
Coated kapok fiber for removal of spilled oil.
Marine pollution bulletin (2013-02-20)
Jintao Wang, Yian Zheng, Aiqin Wang
ABSTRACT
Based on raw kapok fiber, two kinds of oil absorbers with high sorption capacity were prepared by a facile solution-immersion process. The coated polymer with low surface energy and rough fiber surface play important role in the retention of oil. The as-prepared fiber can quickly absorb gasoline, diesel, soybean oil, and paraffin oil up to above 74.5%, 66.8%, 64.4% and 47.8% of oil sorption capacity of raw fiber, respectively. The absorbed oils can be easily recovered by a simple vacuum filtration and the recovered coated-fiber still can be used for several cycles without obvious loss in oil sorption capacity. The thermodynamic study indicates that the adsorption process is spontaneous and exothermic, with complex physisorption and chemisorption. The results suggest that the coated fiber can be used as a low-cost alternative for the removal of oil spilled on water surface.
Sigma-Aldrich
Paraffin wax, mp 53-58 °C (ASTM D 87)
Sigma-Aldrich
Paraffin wax, tested according to Ph. Eur., high viscosity
Sigma-Aldrich
Paraffin wax, chunks (large), white
Supelco
Paraffin oil, suitable for IR spectroscopy
Sigma-Aldrich
Paraffin wax, tested according to Ph. Eur., low viscosity
Sigma-Aldrich
Paraffin wax, meets analytical specification of Ph. Eur., white, pastilles
Sigma-Aldrich
Paraffin wax, mp ≥65 °C
Sigma-Aldrich
Paraffin wax, pellets, white, tested according to Ph. Eur.
Sigma-Aldrich
Paraffin wax, pellets, white
Sigma-Aldrich
Paraffin wax, mp 58-62 °C (ASTM D 87)
Sigma-Aldrich
Paraffin oil, puriss., meets analytical specification of Ph. Eur., BP, viscous liquid
Supelco
Soybean oil, analytical standard
Sigma-Aldrich
Soybean oil, dietary source of long-chain triglycerides and other lipids