Merck
  • Insight into changes during coagulation in NOM reactivity for trihalomethanes and haloacetic acids formation.
Insight into changes during coagulation in NOM reactivity for trihalomethanes and haloacetic acids formation.
Journal of environmental management (2013-02-23)
Aleksandra Tubić, Jasmina Agbaba, Božo Dalmacija, Jelena Molnar, Snežana Maletić, Malcolm Watson, Svetlana Ugarčina Perović
ABSTRACT
Natural organic matter (NOM) in raw water can contribute in many ways to the poor quality of drinking water, including the formation of disinfection byproducts such as trihalomethanes (THM) and haloacetic acids (HAA) during disinfection. This paper investigates the role of individual NOM fractions on changes in THM and HAA formation during coagulation with iron chloride (FeCl3) and a combination of polyaluminium chloride and iron chloride (FeCl3/PACl). The dissolved organic carbon (DOC) in the raw water and after coagulation was fractionated into four fractions, based on their hydrophobicity. Fractionation showed that most of the DOC (68%) in the raw water comes from the fulvic acid fraction, yielding 41% of the total THM precursors and 21% of the total HAA precursors. Both coagulants remove the humic acid fraction, but result in different changes to the reactivity of the remaining NOM fractions towards THM and HAA formation, indicating that coagulation occurs by different pathways, depending upon the type of coagulant used. In particular, significant changes in the reactivities of the hydrophilic acidic and non-acidic fractions were observed.
Sigma-Aldrich
Aluminum hydroxide, reagent grade
Sigma-Aldrich
Iron(II) chloride, 98%
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Iron(II) chloride tetrahydrate, 99.99% trace metals basis
Sigma-Aldrich
Iron(II) chloride tetrahydrate, ReagentPlus®, 98%
Sigma-Aldrich
Iron(II) chloride tetrahydrate, puriss. p.a., ≥99.0% (RT)