Merck
  • Remarkable effect of CF3CH2OH for the halogen induced oxidative rearrangement reaction of aminals leading to 3,4-dihydroquinazolines.
Remarkable effect of CF3CH2OH for the halogen induced oxidative rearrangement reaction of aminals leading to 3,4-dihydroquinazolines.
Organic & biomolecular chemistry (2013-03-06)
Kenichi Murai, Masato Shimura, Ryu Nagao, Daisuke Endo, Hiromichi Fujioka
ABSTRACT
CF3CH2OH was found to be a useful solvent for the oxidative rearrangement reactions of aminals promoted by N-chlorosuccinimide, which proceed via the intermediacy of in situ formed chloro-aminals and that produce 3,4-dihydroquinazolines.
Sigma-Aldrich
2,2,2-Trifluoroethanol, ReagentPlus®, ≥99%
Supelco
2,2,2-Trifluoroethanol, analytical standard, suitable for NMR (spectroscopy)
Sigma-Aldrich
N-Chlorosuccinimide, 98%