Merck
  • Legendre and Laguerre polynomial approach for modeling of wave propagation in layered magneto-electro-elastic media.
Legendre and Laguerre polynomial approach for modeling of wave propagation in layered magneto-electro-elastic media.
The Journal of the Acoustical Society of America (2013-03-08)
Olivier Bou Matar, Noura Gasmi, Huan Zhou, Marc Goueygou, Abdelkrim Talbi
ABSTRACT
A numerical method to compute propagation constants and mode shapes of elastic waves in layered piezoelectric-piezomagnetic composites, potentially deposited on a substrate, is described. The basic feature of the method is the expansion of particle displacement, stress fields, electric and magnetic potentials in each layer on different polynomial bases: Legendre for a layer of finite thickness and Laguerre for the semi-infinite substrate. The exponential convergence rate of the method for propagation of Love waves is numerically verified. The main advantage of the method is to directly determine complex wave numbers for a given frequency via a matricial eigenvalue problem, in a way that no transcendental equation has to be solved. Results are presented and the method is discussed.
Sigma-Aldrich
Barium titanate(IV), powder, <3 μm, 99%
Sigma-Aldrich
Barium titanate(IV), nanopowder (cubic crystalline phase), <100 nm particle size (BET), ≥99% trace metals basis
Sigma-Aldrich
Lithium niobate, 99.9% trace metals basis
Sigma-Aldrich
Barium titanate(IV), powder, <2 μm, 99.5% trace metals basis
Sigma-Aldrich
Niobium, powder, <45 μm, 99.8% trace metals basis
Sigma-Aldrich
Niobium, foil, thickness 0.25 mm, 99.8% trace metals basis
Niobium, IRMM®, certified reference material, 0.02 mm foil
Niobium, IRMM®, certified reference material, 0.5 mm wire
Niobium, IRMM®, certified reference material, 0.5 mm wire
Niobium, IRMM®, certified reference material, 0.02 mm foil