Merck
  • A genome-wide RNAi screen in Caenorhabditis elegans identifies the nicotinic acetylcholine receptor subunit ACR-7 as an antipsychotic drug target.
A genome-wide RNAi screen in Caenorhabditis elegans identifies the nicotinic acetylcholine receptor subunit ACR-7 as an antipsychotic drug target.
PLoS genetics (2013-03-08)
Taixiang Saur, Sarah E DeMarco, Angelica Ortiz, Gregory R Sliwoski, Limin Hao, Xin Wang, Bruce M Cohen, Edgar A Buttner
ABSTRACT
We report a genome-wide RNA interference (RNAi) screen for Suppressors of Clozapine-induced Larval Arrest (scla genes) in Caenorhabditis elegans, the first genetic suppressor screen for antipsychotic drug (APD) targets in an animal. The screen identifies 40 suppressors, including the α-like nicotinic acetylcholine receptor (nAChR) homolog acr-7. We validate the requirement for acr-7 by showing that acr-7 knockout suppresses clozapine-induced larval arrest and that expression of a full-length translational GFP fusion construct rescues this phenotype. nAChR agonists phenocopy the developmental effects of clozapine, while nAChR antagonists partially block these effects. ACR-7 is strongly expressed in the pharynx, and clozapine inhibits pharyngeal pumping. acr-7 knockout and nAChR antagonists suppress clozapine-induced inhibition of pharyngeal pumping. These findings suggest that clozapine activates ACR-7 channels in pharyngeal muscle, leading to tetanus of pharyngeal muscle with consequent larval arrest. No APDs are known to activate nAChRs, but a number of studies indicate that α7-nAChR agonists may prove effective for the treatment of psychosis. α-like nAChR signaling is a mechanism through which clozapine may produce its therapeutic and/or toxic effects in humans, a hypothesis that could be tested following identification of the mammalian ortholog of C. elegans acr-7.
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
Sigma-Aldrich
Clozapine
Clozapine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(±)-Nicotine, ≥99% (TLC), liquid
Supelco
Clozapine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Clozapine for peak identification, European Pharmacopoeia (EP) Reference Standard