Merck
Microbiology of urinary tract infections in Gaborone, Botswana.
PloS one (2013-03-08)
Andrew J Renuart, David M Goldfarb, Margaret Mokomane, Ephraim O Tawanana, Mohan Narasimhamurthy, Andrew P Steenhoff, Jonathan A Silverman
ABSTRACT
The microbiology and epidemiology of UTI pathogens are largely unknown in Botswana, a high prevalence HIV setting. Using laboratory data from the largest referral hospital and a private hospital, we describe the major pathogens causing UTI and their antimicrobial resistance patterns. This retrospective study examined antimicrobial susceptibility data for urine samples collected at Princess Marina Hospital (PMH), Bokamoso Private Hospital (BPH), or one of their affiliated outpatient clinics. A urine sample was included in our dataset if it demonstrated pure growth of a single organism and accompanying antimicrobial susceptibility and subject demographic data were available. A total of 744 samples were included. Greater than 10% resistance was observed for amoxicillin, co-trimoxazole, amoxicillin-clavulanate, and ciprofloxacin. Resistance of E. coli isolates to ampicillin and co-trimoxazole was greater than 60% in all settings. HIV status did not significantly impact the microbiology of UTIs, but did impact antimicrobial resistance to co-trimoxazole. Data suggests that antimicrobial resistance has already emerged to most oral antibiotics, making empiric management of outpatient UTIs challenging. Ampicillin, co-trimoxazole, and ciprofloxacin should not be used as empiric treatment for UTI in this context. Nitrofurantoin could be used for simple cystitis; aminoglycosides for uncomplicated UTI in inpatients.
Product Description
Supelco
Ciprofloxacin HCl, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Nitrofurantoin, VETRANAL®, analytical standard
Supelco
Ampicillin trihydrate, VETRANAL®, analytical standard
Sigma-Aldrich
Nitrofurantoin, 98.0-102.0% (EP, UV), meets EP testing specifications
Sigma-Aldrich
Ampicillin trihydrate, 900-1050 μg/mg anhydrous basis (HPLC)
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Supelco
Nitrofurantoin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Ampicillin sodium salt, BioXtra, suitable for cell culture
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Ciprofloxacin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Ampicillin
Ciprofloxacin, European Pharmacopoeia (EP) Reference Standard
Ciprofloxacin hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Ampicillin sodium salt
Sigma-Aldrich
Ampicillin sodium salt, powder or crystals, BioReagent, suitable for cell culture