Merck
  • Formulation and evaluation of clozapine orally disintegrating tablets prepared by direct compression.
Formulation and evaluation of clozapine orally disintegrating tablets prepared by direct compression.
Die Pharmazie (2013-03-09)
S S Olmez, I Vural, S Sahin, A Ertugrul, Y Capan
ABSTRACT
In this study, clozapine orally disintegrating tablets (ODTs) were prepared by direct compression method. Disintegration time, resistance to crushing of tablets, porosity, friability, dissolution tests were performed and dissolution profiles of ODTs were investigated. Morphological and interaction studies were also performed. Friability values were found to be less than 1%. All tablet formulations disintegrated within 1 min and fulfilled the 3 min disintegration time required for ODTs given in the European Pharmacopoeia. More than 85% of the labeled amount of clozapine was dissolved in 15 min from the ODTs. No interaction or changes were found between active substance and excipients. As a result of the studies, ODT formulations developed in this study can be suggested as promising formulations, which assist development and manufacturing a generic product of clozapine.
Product Description
Supelco
Povidone, Pharmaceutical Secondary Standard; Certified Reference Material
Clozapine, European Pharmacopoeia (EP) Reference Standard
Povidone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Starch from potato, Soluble
Sigma-Aldrich
Starch from wheat, Unmodified
Sigma-Aldrich
Starch from potato, Powder
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 1.2
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.7
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.9
Sigma-Aldrich
Sodium carboxymethyl cellulose, viscosity 50-200 cP , c=4% H2O at 25­°C
Sigma-Aldrich
Sodium carboxymethyl cellulose, viscosity 2500-6000 cP 
Sigma-Aldrich
Carboxymethylcellulose sodium, meets USP testing specifications, Medium viscosity
Sigma-Aldrich
Povidone, meets USP testing specifications
Sigma-Aldrich
Kollidon® 25
Sigma-Aldrich
Clozapine
Sigma-Aldrich
Polyvinylpyrrolidone, K 30
Sigma-Aldrich
Carboxymethylcellulose sodium salt, low viscosity
Supelco
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10
Supelco
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
Sigma-Aldrich
Polyvinylpyrrolidone, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Carboxymethylcellulose sodium salt, ultra high viscosity, highly purified
Sigma-Aldrich
Carboxymethylcellulose sodium salt, Medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, High viscosity
Supelco
Clozapine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Starch from corn, practical grade
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.
Sigma-Aldrich
Starch from corn