Merck
  • Pharmacogenetics of CYP1A2 activity and inducibility in smokers and exsmokers.
Pharmacogenetics of CYP1A2 activity and inducibility in smokers and exsmokers.
Pharmacogenetics and genomics (2013-03-16)
Maria Dobrinas, Jacques Cornuz, Chin B Eap
ABSTRACT
There is a high interindividual variability in cytochrome P4501A2 (CYP1A2) activity and in its inducibility by smoking, only poorly explained by known CYP1A2 polymorphisms. We aimed to study the contribution of other regulatory pathways, including transcription factors and nuclear receptors, toward this variability. CYP1A2 activity was determined by the paraxanthine/caffeine ratio in 184 smokers and in 113 of them who were abstinent for 4 weeks. Participants were genotyped for 22 polymorphisms in 12 genes. A significant influence on CYP1A2 inducibility was observed for the NR1I3 rs2502815 (P=0.0026), rs4073054 (P=0.029), NR2B1 rs3818740 (P=0.0045), rs3132297 (P=0.036), AhR rs2282885 (P=0.040), rs2066853 (P=0.019), NR1I1 rs2228570 (P=0.037), and NR1I2 rs1523130 (P=0.044) polymorphisms. Among these, the NR1I3 rs2502815 (P=0.0045), rs4073054 (P=0.048), and NR2B1 rs3818740 (P=0.031) also influenced CYP1A2 basal activity. This is the first in-vivo demonstration of the influence of genes involved in CYP1A2 regulatory pathways on its basal activity and inducibility by smoking. These results need to be confirmed by other studies.
Product Description
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Caffeine, European Pharmacopoeia (EP) Reference Standard
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®