Merck
[Effect of anticonvulsants on the nitric oxide system].
Ukrains'kyi biokhimichnyi zhurnal (1999 ) (2013-03-29)
L A Gromov, I F Belenichev, L G Gonchar-Cherdakli, G A Zhernovaia
ABSTRACT
The effect of phenobarbital, carbamazepine, valproate sodium, depakine, topiramate and lamotrigine on the content of NO and NO-synthase activity in white rat brain tissues has been studied. It was established that the action of carbamazepine, valproate sodium, topiramate and lamotrigine decreases the activity of NO-synthase and the level of NO in the brain tissues. The amount of NO does not change while NO-synthase activity increases with the introduction of phenobarbital. The involvement of nitric oxide in the mechanism of action of the studied anticonvulsant drugs is discussed.
Product Description
Supelco
Valproic acid solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Carbamazepine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Carbamazepine, powder
Supelco
Phenobarbital solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
2-Propylpentanoic acid
Supelco
Valproic acid, Pharmaceutical Secondary Standard; Certified Reference Material