Merck
  • Permethrin resistance in Aedes albopictus (Diptera: Culicidae) and associated fitness costs.
Permethrin resistance in Aedes albopictus (Diptera: Culicidae) and associated fitness costs.
Journal of medical entomology (2013-04-02)
Hiang Hao Chan, Jaal Zairi
ABSTRACT
Insecticide resistance has become a serious issue in vector management programs. Information on insecticidal resistance and its associated mechanisms is important for successful insecticide resistance management. The selection of a colony of permethrin-resistant Aedes albopictus (Skuse) (Diptera: Culicidae), originating from Penang Island, Malaysia, yielded high larval-specific resistance to permethrin and cross-resistance to deltamethrin. Synergism assays showed that the major mechanism underlying this resistance involves cytochrome P450 monooxygenase. The resistance is autosomal, polygenically inherited and incompletely dominant (D = 0.26). Resistant larvae were reared under different conditions to assess the fitness costs. Under high larval density, larval development time of the resistant SGI strain was significantly longer than the susceptible VCRU strain. In both high- and low-density conditions SGI showed a lower rate of emergence and survival compared with the VCRU strain. Resistant larvae were more susceptible to predation by Toxorhynchites splendens (Wiedemann) (Diptera: Culicidae) larvae. The body size of SGI females reared under high-density conditions was larger compared with females of the susceptible strain. SGI females survived longer when starved than did VCRU females. The energy reserve upon eclosion was positively correlated with the size of the adults.
Product Description
Supelco
Permethrin, PESTANAL®, analytical standard