Merck
Oxcarbazepine for neuropathic pain.
The Cochrane database of systematic reviews (2013-04-02)
Muke Zhou, Ning Chen, Li He, Mi Yang, Cairong Zhu, Fengbo Wu
ABSTRACT
Neuropathic pain is difficult to treat. Oxcarbazepine is an anticonvulsant drug closely related to carbamazepine and is reportedly better tolerated. Oxcarbazepine has been reported to be efficacious in the treatment of neuropathic pain. To determine the benefits and harms of oxcarbazepine for different forms of neuropathic pain. We searched the Cochrane Neuromuscular Disease Group Specialized Register (30 October 2012), CENTRAL (2012, Issue 10), MEDLINE (January 1966 to October 2012), EMBASE (January 1980 to October 2012) and the Chinese Biomedical Retrieval System (January 1978 to October 2012) for trials. We also searched the National Institutes of Health (NIH) databases and the World Health Organization (WHO) International Clinical Trials Registry Platform for ongoing trials, and wrote to the companies who make oxcarbazepine and to pain experts asking for additional information. All randomised controlled trials and cross-over studies of oxcarbazepine for the treatment of people of any age or sex with any neuropathic pain were eligible. We planned to include trials of oxcarbazepine compared with placebo or any other intervention, regardless of administration route, dosage or length of treatment. Two review authors independently selected the studies for inclusion, assessed their risk of bias, extracted data and typed the data onto forms. The authors resolved any disagreements through discussion. Four multicentre, randomised, placebo-controlled, double-blind trials with a total of 779 participants were eligible for inclusion. These were from a series of studies funded by the manufacturer. Three of them investigated oxcarbazepine in people with painful diabetic neuropathy (634 participants) and one was a trial of oxcarbazepine for neuropathic pain due to radiculopathy (145 participants). Although these trials were well designed, the imbalanced and large amount of incomplete outcome data led to a risk of bias in the results. Results for painful diabetic neuropathy showed that compared to the baseline the proportion of participants who reported a 50% or 30% reduction of pain scores after 16 weeks of treatment was significantly higher in the oxcarbazepine group than the placebo group (50% reduction: risk ratio (RR) 1.91, 95% confidence interval (CI) 1.08 to 3.39, number of people needed to treat for an additional beneficial outcome (NNTB) 6.0, 95% CI 3.3 to 41.0; 30% reduction: RR 1.57, 95% CI 1.01 to 2.44, NNTB 6.1, 95% CI 3.1 to 113.6). However, both results were based on data from the single positive trial (146 participants) since the two negative trials did not provide data that could be included in a meta-analysis. For participants with neuropathic pain due to radiculopathy, the trial demonstrated no significant efficacy for oxcarbazepine. Although trial reports stated that most side effects were mild to moderate in severity, the proportion of events leading to withdrawals was statistically higher in the oxcarbazepine group than in the placebo group both for painful diabetic neuropathy (RR 3.86, 95% CI 2.29 to 6.40) and radiculopathy (RR 2.84, 95% CI 1.55 to 5.23). On the basis of moderate quality evidence from one trial in diabetic peripheral neuropathy, oxcarbazepine is effective in reducing pain for this condition. However, this conclusion does not take into account negative results from other trials in diabetic peripheral neuropathy that could not be included in our meta-analysis. We did not find any evidence from randomised controlled trials to determine the efficacy or safety of oxcarbazepine for other kinds of neuropathic pain. Most adverse effects related to oxcarbazepine are rated as mild to moderate in severity, but adverse events leading to discontinuation of drug administration or serious adverse events are not uncommon. More well designed randomised controlled trials investigating oxcarbazepine for various types of neuropathic pain are needed.
Product Description
Supelco
Carbamazepine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
USP
Oxcarbazepine, United States Pharmacopeia (USP) Reference Standard
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
Oxcarbazepine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Carbamazepine, powder
Supelco
Oxcarbazepine solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Oxcarbazepine, ≥98% (HPLC), solid