Merck
  • Pharmacokinetics of a new immediate-release compound omeprazole capsule and its comparison with the enteric-coated formulation under fasting and fed conditions.
Pharmacokinetics of a new immediate-release compound omeprazole capsule and its comparison with the enteric-coated formulation under fasting and fed conditions.
Drug research (2013-04-06)
Z Liu, L Ding, S Zhong, X Cao, L Jiang, H Duan
ABSTRACT
The present study was conducted to describe the pharmacokinetic profile of immediate-release compound omeprazole capsule and compare it with the enteric-coated formulation under fasting and fed condition.This study was designed to phase I, open-label, randomized, 3-part clinical trial. 12 subjects in part one received single doses (20 mg and 40 mg) and repeated doses (20 mg). Different 30 subjects in part 2 (fasting condition) and part 3 (fed condition) received either compound omeprazole capsule (40 mg) or delayed-release omeprazole (40 mg) separated by 7 days.Compound omeprazole capsule showed dose non-proportionality in the range from 20 mg to 40 mg. The AUC0-t of 20 mg omeprazole is 78% higher after repeated doses. The C max was higher and T max was lower for compound omeprazole capsule than delayed-release omeprazole. 90% CIs for AUC0-t of the 2 periods (test/reference) under fasting and fed conditions were 106.3% (102.2%~109.2%) and 104.1% (93.4%~109.6%), respectively. Presence of food reduced the rate (C max) and extent (AUC0-t) of systemic exposure of the test and reference formulations from 1 462 to 777 ng · mL-1, 1 055 to 602 ng · mL-1, 2 597 to 1 852 ng · h · mL-1 and 2 454 to 1 873 ng · h · mL-1, respectively.Exposed dose of omeprazole in subjects with compound omeprazole capsule was time- and dose-dependent. Compared with delayed-release omeprazole, compound omeprazole capsule had rapid but similar degree of absorption of omeprazole.
Product Description
Supelco
Omeprazole, Pharmaceutical Secondary Standard; Certified Reference Material
Omeprazole for peak identification, European Pharmacopoeia (EP) Reference Standard
Omeprazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Omeprazole, solid