Merck
  • Evidence that a metabolic microcompartment contains and recycles private cofactor pools.
Evidence that a metabolic microcompartment contains and recycles private cofactor pools.
Journal of bacteriology (2013-04-16)
Douglas L Huseby, John R Roth
ABSTRACT
Microcompartments are loose protein cages that encapsulate enzymes for particular bacterial metabolic pathways. These structures are thought to retain and perhaps concentrate pools of small, uncharged intermediates that would otherwise diffuse from the cell. In Salmonella enterica, a microcompartment encloses enzymes for ethanolamine catabolism. The cage has been thought to retain the volatile intermediate acetaldehyde but allow diffusion of the much larger cofactors NAD and coenzyme A (CoA). Genetic tests support an alternative idea that the microcompartment contains and recycles private pools of the large cofactors NAD and CoA. Two central enzymes convert ethanolamine to acetaldehyde (EutBC) and then to acetyl-CoA (EutE). Two seemingly peripheral redundant enzymes encoded by the eut operon proved to be essential for ethanolamine utilization, when subjected to sufficiently stringent tests. These are EutD (acetyl-CoA to acetyl phosphate) and EutG (acetaldehyde to ethanol). Obligatory recycling of cofactors couples the three reactions and drives acetaldehyde consumption. Loss and toxic effects of acetaldehyde are minimized by accelerating its consumption. In a eutD mutant, acetyl-CoA cannot escape the compartment but is released by mutations that disrupt the structure. The model predicts that EutBC (ethanolamine-ammonia lyase) lies outside the compartment, using external coenzyme B12 and injecting its product, acetaldehyde, into the lumen, where it is degraded by the EutE, EutD, and EutG enzymes using private pools of CoA and NAD. The compartment appears to allow free diffusion of the intermediates ethanol and acetyl-PO4 but (to our great surprise) restricts diffusion of acetaldehyde.
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Coenzyme A hydrate, ≥85% (UV, HPLC)
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Supelco
Ethanolamine, analytical standard
Sigma-Aldrich
Ethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC/NT)
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Ethanolamine, ≥99%
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethanolamine, ≥98%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥95% (HPLC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, Grade AA-1
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, purified by column chromatography, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥98%, BioUltra, from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥99%
Supelco
Acetaldehyde, PESTANAL®, analytical standard
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetaldehyde, natural, FG
Sigma-Aldrich
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)