Merck
  • Heme degradation by Staphylococcus aureus IsdG and IsdI liberates formaldehyde rather than carbon monoxide.
Heme degradation by Staphylococcus aureus IsdG and IsdI liberates formaldehyde rather than carbon monoxide.
Biochemistry (2013-04-23)
Toshitaka Matsui, Shusuke Nambu, Yukari Ono, Celia W Goulding, Kouhei Tsumoto, Masao Ikeda-Saito
ABSTRACT
IsdG and IsdI from Staphylococcus aureus are novel heme-degrading enzymes containing unusually nonplanar (ruffled) heme. While canonical heme-degrading enzymes, heme oxygenases, catalyze heme degradation coupled with the release of CO, in this study we demonstrate that the primary C1 product of the S. aureus enzymes is formaldehyde. This finding clearly reveals that both IsdG and IsdI degrade heme by an unusual mechanism distinct from the well-characterized heme oxygenase mechanism as recently proposed for MhuD from Mycobacterium tuberculosis. We conclude that heme ruffling is critical for the drastic mechanistic change for these novel bacterial enzymes.
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O