Merck
  • [Diagnosis of non dermatophyte onychomycosis ant its relevance for treatment].
[Diagnosis of non dermatophyte onychomycosis ant its relevance for treatment].
Revue medicale suisse (2013-04-30)
Michel Monod, Massimo Lurati, Florence Baudraz-Rosselet
ABSTRACT
PCR methods are reliable and suitable to in situ identify dermatophytes, yeasts and non dermatophyte moulds (NDM) in onychomycosis. Onychomycosis insensitive to standard treatment with topical agents as well as with oral terbinafine or itraconazole revealed Fusarium spp., Acremonium spp. and Aspergillus spp. as infectious agents. However, NDM onychomycosis could be efficiently cured using topical amphotericin B. In conclusion, correct fungal species identification is important in onychomycoses in order to prescribe adequate treatments since dermatophytes and moulds have different sensitivities to antifungal drugs.
Sigma-Aldrich
Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture
Amphotericin B for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Amphotericin B from Streptomyces sp., BioReagent, suitable for cell culture, ~80% (HPLC)
Sigma-Aldrich
Amphotericin B from Streptomyces sp., ~80% (HPLC), powder
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture