Merck
  • Facile synthesis of multifunctional beta-NaGdF4:Yb3+/Er3+ nanoparticles in oleylamine.
Facile synthesis of multifunctional beta-NaGdF4:Yb3+/Er3+ nanoparticles in oleylamine.
Journal of nanoscience and nanotechnology (2013-05-08)
Qian Cheng, Jiehe Sui, Yu Li, Ziqiao Zhou, Wei Cai
ABSTRACT
Multifunctional beta-NaGdF4:Yb3+/Er3+ nanoparticles (NPs) were successfully synthesized using oleylamine as both solvent and stabilizer via the thermolysis method. They have uniform morphology with a mean size of 12.7 nm and show efficient up-conversion emission when excited by a 980 nm laser. The up-conversion NPs demonstrated a nearly quadratic dependence of the photoluminescence intensity on the excitation power, which indicated a two-photon induced process. In addition, these NPs exhibit paramagnetic characteristics at both 300 and 77 K. The magnetic properties of beta-NaGdF4:Yb3+/Er3+ NPs are intrinsic to the Gd3+ ions. The measured mass magnetic susceptibility value of 0.79 x 10(-4) em mu/g x Oe at room temperature is close to reported values of other NPs for bioseparation and optical-magnetic dual modal nanoprobes in biomedical imaging.
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Gadolinium, −40 mesh, 99% trace rare earth metals basis
Sigma-Aldrich
Gadolinium, chips
Sigma-Aldrich
Oleylamine, technical grade, 70%
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%