Merck
  • Nanofibrous rutile-titania/graphite composite derived from natural cellulose substance.
Nanofibrous rutile-titania/graphite composite derived from natural cellulose substance.
Journal of nanoscience and nanotechnology (2013-05-08)
Yan Luo, Xiaoyan Liu, Jianguo Huang
ABSTRACT
Calcination and carbonization of ultrathin titania gel film pre-coated cellulose nanofibers of ordinary laboratory filter paper at 1300 degrees C under argon atmosphere yielded a hierarchical nanofibrous rutile-titania/carbon composite material. The carbon fibers derived from the initial cellulose nanofibers possess a core-shell structure with amorphous carbon at the inner part and graphite state carbon at the outer part. The initial titania gel films were converted into rutile phase titania nanoparticles with sizes of 20-50 nm anchored on the surface of the carbon fibers. The resulting composite possesses considerable electric conductivity as well as decolorization activity towards dye-bearing wastewater.
Product Description
Supelco
Cellulose, acid washed, from spruce, suitable for column chromatography
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Sigma-Aldrich
Cellulose, fibers, (medium)
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Supelco
Cellulose, acid washed, powder, suitable for column chromatography
Sigma-Aldrich
Cellulose, microcrystalline, powder
Supelco
Cellulose, powder, suitable for column chromatography
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm
Sigma-Aldrich
Cellulose, colloidal, microcrystalline
Sigma-Aldrich
Graphite, powder, <150 μm, 99.99% trace metals basis
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
α-Cellulose, powder
Sigma-Aldrich
Graphite, powder, <20 μm, synthetic
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 3 mm, low density, 99.995% trace metals basis
Sigma-Aldrich
Graphite, flakes
Sigma-Aldrich
Graphite, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 6 mm, 99.995% trace metals basis