Merck
  • [Interaction of surface-active base with fraction of membrane-bound Williams's protons].
[Interaction of surface-active base with fraction of membrane-bound Williams's protons].
Biofizika (2013-05-09)
L S Iaguzhinskiĭ, K A Motovilov, E M Volkov, S A Eremeev
ABSTRACT
In the process of mitochondrial respiratory H(+)-pumps functioning, the fraction membrane-bound protons (R-protons), which have an excess of free energy is formed. According to R.J. Williams this fraction is included as energy source in the reaction of ATP synthesis. Previously, in our laboratory was found the formation of this fraction was found in the mitochondria and on the outer surface of mitoplast. On the mitoslast model we strictly shown that non-equilibrium R-proton fraction is localized on the surface of the inner mitochondrial membrane. In this paper a surface-active compound--anion of 2,4,6-trichloro-3-pentadecylphenol (TCP-C15) is described, which selectively interacts with the R-protons fraction in mitochondria. A detailed description of the specific interaction of the TCP-C15 with R-protons fraction in mitochondria is presented. Moreover, in this work it was found that phosphate transport system reacts with the R-protons fraction in mitochondria and plays the role of the endogenous volume regulation system of this fraction. The results of experiments are discussed in the terms of a local coupling model of the phosphorylation mechanism.
Sigma-Aldrich
2,4,6-Trichlorophenol, 98%