Merck
  • An Arabidopsis R2R3-MYB transcription factor, AtMYB20, negatively regulates type 2C serine/threonine protein phosphatases to enhance salt tolerance.
An Arabidopsis R2R3-MYB transcription factor, AtMYB20, negatively regulates type 2C serine/threonine protein phosphatases to enhance salt tolerance.
FEBS letters (2013-05-11)
Mei Hua Cui, Kyoung Shin Yoo, Sujin Hyoung, Ha Thi Kim Nguyen, Yun Young Kim, Hae Jin Kim, Sung Han Ok, Sang Dong Yoo, Jeong Sheop Shin
ABSTRACT
We have characterized the function of a plant R2R3-MYB transcription factor, Arabidopsis thaliana MYB20 (AtMYB20). Transgenic plants overexpressing AtMYB20 (AtMYB20-OX) enhanced salt stress tolerance while repression lines (AtMYB20-SRDX) were more vulnerable to NaCl than wild-type plants. Following NaCl treatment, the expressions of ABI1, ABI2 and AtPP2CA, which encode type 2C serine/threonine protein phosphatases (PP2Cs) that act as negative regulators in abscisic acid (ABA) signaling, were suppressed in AtMYB20-OX but induced in AtMYB20-SRDX. The electrophoretic mobility shift assay results revealed that AtMYB20 binds to the promoter regions containing the MYB recognition sequence (TAACTG) and an ACGT core element of ABI1 and AtPP2CA. These findings suggest that AtMYB20 down-regulates the expression of PP2Cs, the negative regulator of ABA signaling, and enhances salt tolerance.
Sigma-Aldrich
2-cis,4-trans-Abscisic acid, synthetic, 98%
Sigma-Aldrich
(+)-Abscisic acid, ≥98% (HPLC)