Merck
  • Robo3-driven axon midline crossing conditions functional maturation of a large commissural synapse.
Robo3-driven axon midline crossing conditions functional maturation of a large commissural synapse.
Neuron (2013-05-15)
Nicolas Michalski, Norbert Babai, Nicolas Renier, David J Perkel, Alain Chédotal, Ralf Schneggenburger
ABSTRACT
During the formation of neuronal circuits, axon pathfinding decisions specify the location of synapses on the correct brain side and in correct target areas. We investigated a possible link between axon midline crossing and the subsequent development of output synapses formed by these axons. Conditional knockout of Robo3 in the auditory system forced a large commissural synapse, the calyx of Held, to be exclusively formed on the wrong, ipsilateral side. Ipsilateral calyx of Held synapses showed strong transmission defects, with reduced and desynchronized transmitter release, fewer fast-releasable vesicles, and smaller and more variable presynaptic Ca(2+) currents. Transmission defects were not observed in a downstream inhibitory synapse, and some defects persisted into adulthood. These results suggest that axon midline crossing conditions functional maturation of commissural synapses, thereby minimizing the impact of mislocalized synapses on information processing. This mechanism might be relevant to human disease caused by mutations in the ROBO3 gene.
Sigma-Aldrich
Cesium, ingot, ≥99.95% trace metals basis
Sigma-Aldrich
Cesium chloride, Grade I, ≥99.0%
Sigma-Aldrich
Cesium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Cesium chloride, 99.99% trace metals basis
Sigma-Aldrich
Cesium chloride, ≥99.999% trace metals basis
Sigma-Aldrich
Cesium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Cesium chloride, Grade II, ≥98%
Sigma-Aldrich
Cesium chloride, Molecular Biology, ≥98%
Sigma-Aldrich
Cesium chloride, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
Cesium chloride, optical grade, ≥99.5% trace metals basis
Sigma-Aldrich
Cesium chloride, Molecular Biology, ≥99% (silver nitrate titration)
Sigma-Aldrich
Cesium chloride, ReagentPlus®, 99.9%
Sigma-Aldrich
Cesium chloride, puriss. p.a., ≥99.5%