Merck
  • Lead optimization of a 4-aminopyridine benzamide scaffold to identify potent, selective, and orally bioavailable TYK2 inhibitors.
Lead optimization of a 4-aminopyridine benzamide scaffold to identify potent, selective, and orally bioavailable TYK2 inhibitors.
Journal of medicinal chemistry (2013-05-15)
Jun Liang, Anne van Abbema, Mercedesz Balazs, Kathy Barrett, Leo Berezhkovsky, Wade Blair, Christine Chang, Donnie Delarosa, Jason DeVoss, Jim Driscoll, Charles Eigenbrot, Nico Ghilardi, Paul Gibbons, Jason Halladay, Adam Johnson, Pawan Bir Kohli, Yingjie Lai, Yanzhou Liu, Joseph Lyssikatos, Priscilla Mantik, Kapil Menghrajani, Jeremy Murray, Ivan Peng, Amy Sambrone, Steven Shia, Young Shin, Jan Smith, Sue Sohn, Vickie Tsui, Mark Ultsch, Lawren C Wu, Yisong Xiao, Wenqian Yang, Judy Young, Birong Zhang, Bing-yan Zhu, Steven Magnuson
ABSTRACT
Herein we report our lead optimization effort to identify potent, selective, and orally bioavailable TYK2 inhibitors, starting with lead molecule 3. We used structure-based design to discover 2,6-dichloro-4-cyanophenyl and (1R,2R)-2-fluorocyclopropylamide modifications, each of which exhibited improved TYK2 potency and JAK1 and JAK2 selectivity relative to 3. Further optimization eventually led to compound 37 that showed good TYK2 enzyme and interleukin-12 (IL-12) cell potency, as well as acceptable cellular JAK1 and JAK2 selectivity and excellent oral exposure in mice. When tested in a mouse IL-12 PK/PD model, compound 37 showed statistically significant knockdown of cytokine interferon-γ (IFNγ), suggesting that selective inhibition of TYK2 kinase activity might be sufficient to block the IL-12 pathway in vivo.
Sigma-Aldrich
4-Aminopyridine, ≥99%
Sigma-Aldrich
4-Aminopyridine, 98%
Supelco
Discovery® Cyano Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm