Merck
  • Effect of nine plant volatiles in the field on the sex pheromones of Leguminivora glycinivorella.
Effect of nine plant volatiles in the field on the sex pheromones of Leguminivora glycinivorella.
Natural product communications (2013-05-18)
Daihua Hu, Juntao Feng, Zhihui Wang, Hua Wu, Xing Zhang
ABSTRACT
Attraction of the soybean pod borer, Leguminivora glycinivorella (Matsumura), an economically important pest of soybean, to nine plant volatiles, alone or combined with two kinds of synthetic sex pheromone, ((E,E)-8,10-dodecadienyl acetate (EE8,10-12:Ac), or a blend of EE8,10-12:Ac and (E)-10-dodecenyl acetate in a 10:1 ratio), was evaluated in field trapping experiments in a soybean field in Harbin, China. By themselves, the plant volatiles (dose) linalool (0.1 mg), (Z)-3-hexenyl acetate (0.1 mg), and geraniol (0.1 mg, 1.0 mg) were weakly attractive to L. glycinivorella males, but significantly reduced mean catches when higher doses were combined with pheromones. Conversely, (E)-2-hexenal, benzaldehyde, and phenylacetaldehyde were not attractive to L. glycinivorella males at any dose tested, but significantly increased mean catch when certain doses were combined with the binary pheromone blend. Other plant volatiles, such as (Z)-3-hexen-1-ol, (Z)-3-hexenyl acetate, and (E)-2-hexenyl acetate, were unattractive on their own, but significantly reduced mean catch of L. glycinivorella males when certain doses were combined with the pheromones. These results suggest that efficacy of pheromone-baited traps for survey and monitoring of L. glycinivorella male moths may be enhanced by the addition of specific plant volatiles and that the relative dose is critical.
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Phenylacetaldehyde, ≥90%
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Phenylacetaldehyde, ≥95%, FCC, FG
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Benzaldehyde, European Pharmacopoeia (EP) Reference Standard
Supelco
Benzaldehyde, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzaldehyde, ≥98%, FG, FCC
Sigma-Aldrich
Benzaldehyde, natural, FCC, FG
Sigma-Aldrich
trans-3-Hexen-1-ol, ≥95%, stabilized
Sigma-Aldrich
Benzaldehyde, purified by redistillation, ≥99.5%
Sigma-Aldrich
Benzaldehyde, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzaldehyde, puriss. p.a., ≥99.0% (GC)
Supelco
Benzaldehyde, analytical standard
Sigma-Aldrich
trans-3-Hexen-1-ol, 97%
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
cis-3-Hexenyl acetate, ≥98%, stabilized, FCC, FG
Supelco
cis-3-Hexen-1-ol, analytical standard
Sigma-Aldrich
cis-3-Hexen-1-ol, natural, >98%, FCC, FG
Sigma-Aldrich
cis-3-Hexen-1-ol, 98%
Sigma-Aldrich
Acetaldehyde, natural, FG
Supelco
Acetaldehyde, PESTANAL®, analytical standard
Sigma-Aldrich
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)
Sigma-Aldrich
trans-2-Hexen-1-al, 98%
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Sigma-Aldrich
trans-2-Hexen-1-al, ≥95%, FCC, FG