Merck
  • Mechanism of enhanced optical second-harmonic generation in the conducting pyrochlore-type Pb2Ir2O7-x oxide compound.
Mechanism of enhanced optical second-harmonic generation in the conducting pyrochlore-type Pb2Ir2O7-x oxide compound.
Physical review letters (2013-05-21)
Yasuyuki Hirata, Makoto Nakajima, Yusuke Nomura, Hiroyuki Tajima, Yoshitaka Matsushita, Keiko Asoh, Yoko Kiuchi, Adolfo G Eguiluz, Ryotaro Arita, Tohru Suemoto, Kenya Ohgushi
ABSTRACT
The structural, electronic, and optical properties of pyrochlore-type Pb(2)Ir(2)O(6)O(0.55)('), which is a metal without spatial inversion symmetry at room temperature, were investigated. Structural analysis revealed that the structural distortion relevant to the breakdown of the inversion symmetry is dominated by the Pb-O' network but is very small in the Ir-O network. At the same time, gigantic second-harmonic generation signals were observed, which can only occur if the local environment of the Ir 5d electrons features broken inversion symmetry. First-principles electronic structure calculations reveal that the underlying mechanism for this phenomenon is the induction of the noncentrosymmetricity in the Ir 5d bands by the strong hybridization with O' 2p orbitals. Our results stimulate theoretical study of inversion-broken iridates, where exotic quantum states such as a topological insulator and Dirac semimetal are anticipated.
Product Description
Niobium, IRMM®, certified reference material, 0.5 mm wire
Niobium, IRMM®, certified reference material, 0.5 mm wire
Niobium, IRMM®, certified reference material, 0.02 mm foil
Sigma-Aldrich
Iridium, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Iridium, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.9% trace metals basis
Niobium, IRMM®, certified reference material, 0.02 mm foil
Sigma-Aldrich
Niobium, powder, <45 μm, 99.8% trace metals basis
Sigma-Aldrich
Iridium, powder, 99.9% trace metals basis
Sigma-Aldrich
Lead, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Niobium, foil, thickness 0.25 mm, 99.8% trace metals basis
Sigma-Aldrich
Iridium(IV) oxide, 99.9% trace metals basis
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis