Merck
  • Tumor cell-derived exosome-targeted dendritic cells stimulate stronger CD8+ CTL responses and antitumor immunities.
Tumor cell-derived exosome-targeted dendritic cells stimulate stronger CD8+ CTL responses and antitumor immunities.
Biochemical and biophysical research communications (2013-05-28)
Ye Yao, Linjun Chen, Wei Wei, Xiaohui Deng, Liyuan Ma, Siguo Hao
ABSTRACT
Tumor cell-derived exosomes (TEX) have been widely used to induce antitumor immune responses in animal models and clinical trials. However, the efficiency of the antitumor immunity that is induced by TEX is still relatively weak. In this study, we compared the antitumor immunities between EG7 tumor cell-derived exosomes (EXO(EG7)) and EXO(EG7)-targeted dendritic cells (DC(EXO)). We found that EXO(EG7) harbored OVA and peptide major histocompatibility complex I (pMHC-I), which were expressed on its parental EG7 tmor cells, and they could transfer OVA and pMHC-I to dendritic cells (DCs) in vitro. DC(EXO) could more efficiently induce antitumor immunity than EXO(EG7). In addition, we showed that the immune stimulatory effects of EXO(EG7) were dependent on the host DCs and, whereas those of DC(EXO) were not, indicating the important role of the host DCs in TEX vaccines. Taken together, TEX-targeted DCs may be more effective for EXO-based vaccines for the induction of antitumor immunity.
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Biotin, meets USP testing specifications
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Sigma-Aldrich
Biotin, ≥99.0% (T)
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material
Biotin, European Pharmacopoeia (EP) Reference Standard