Merck
  • The formulation of flurbiprofen loaded microspheres using hydroxypropylmethycellulose and ethylcellulose.
The formulation of flurbiprofen loaded microspheres using hydroxypropylmethycellulose and ethylcellulose.
Advances in clinical and experimental medicine : official organ Wroclaw Medical University (2013-05-28)
Muhammad K Shahzad, Muhammad Ubaid, Moosa Raza, Ghulam Murtaza
ABSTRACT
The aim of the present work was to formulate flurbiprofen (FLB) loaded microspheres of hydroxypropylmethycellulose and ethylcellulose polymers to study the effect of different proportions of the polymer mixture on the release behavior of the drug. A series of microspheres were prepared using tween-80 as a surfactant. The prepared microspheres were evaluated for entrapment efficiency (%) and percentage recovery. Drug release was performed in USP phosphate buffers of pH 1.2 and 6.8. Drug release data were plotted in various kinetic models, including zero-order, first-order, Higuchi and Korsmeyer-Peppas models to investigate the optimum composition suitable for sustained drug delivery. A significant difference in drug release kinetics was observed by varying the composition of hydroxypropylmethycellulose/ethylcellulose. As the ratio of EC/HPMC was increased, the release rate of flurbiprofen decreased. This study demonstrated the potential of polymer combinations in the formulation of microspheres for water-insoluble drugs utilizing HPMC and EC as release retardant materials, using a simple solvent evaporation microencapsulation technique. It was observed that various physico-chemical properties of the microspheres varied according to the change in polymer concentrations used in the formulations.
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~120,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~86,000
Sigma-Aldrich
Methyl cellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose
Sigma-Aldrich
Methyl cellulose, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
Methyl cellulose, tested according to Ph. Eur.
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~90,000
Sigma-Aldrich
Methyl cellulose, meets USP testing specifications, 26.0-33.0% (methoxyl group, on Dry Basis), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP
Sigma-Aldrich
Hypromellose, meets USP testing specifications
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
α-Cellulose, powder
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% methoxyl basis
Sigma-Aldrich
Ethyl cellulose, viscosity 100 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethyl cellulose, viscosity 4 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethyl cellulose, viscosity 22 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl