Merck
  • Is a dexamethasone-sparing strategy capable of preventing acute and delayed emesis caused by combined doxorubicin and paclitaxel for breast cancer? Analysis of a phase II trial.
Is a dexamethasone-sparing strategy capable of preventing acute and delayed emesis caused by combined doxorubicin and paclitaxel for breast cancer? Analysis of a phase II trial.
Oncology (2013-05-29)
Silvia Damian, Luigi Celio, Elena De Benedictis, Paola Mariani, Francesco Agustoni, Francesca Ricchini, Filippo De Braud
ABSTRACT
The effectiveness of palonosetron without delayed dexamethasone dosing against emesis was investigated in patients scheduled to receive the corticosteroid-containing combination of doxorubicin and paclitaxel (AT) for 3 cycles. Chemo-naïve women with breast cancer receiving doxorubicin (60 mg/m(2)) and paclitaxel (200 mg/m(2)) were eligible. Patients received palonosetron 0.25 mg intravenously before chemotherapy, however, all patients also received a premedication consisting of prednisone (25 mg orally the evening before therapy) and hydrocortisone (250 mg intravenously just before paclitaxel). The primary end point was complete control (CC; no vomiting, no rescue anti-emetics, and no more than mild nausea) during the overall phase (days 1-5) following cycle 1. Seventy-six patients were enrolled and evaluable (median age 50 years). Fifty-six patients (74%; 95% CI 62-83%) achieved overall CC. Acute (day 1) and delayed (days 2-5) CC rates were 78 and 74%, respectively. No vomiting rates for the acute, delayed and overall phases were 85, 85 and 83%, respectively. An exploratory analysis showed only a small decrease in the probability of achieving CC between cycle 1 (74%) and cycle 3 (66%). The dexamethasone-sparing strategy prevented emesis in more than 70% of breast cancer patients receiving their initial cycle of AT chemotherapy.
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Doxorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Paclitaxel semi-synthetic for peak identification, European Pharmacopoeia (EP) Reference Standard
Paclitaxel, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Paclitaxel semi-synthetic for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone-Water Soluble, BioReagent, suitable for cell culture
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%