Merck
  • Metabolic signatures of amyotrophic lateral sclerosis reveal insights into disease pathogenesis.
Metabolic signatures of amyotrophic lateral sclerosis reveal insights into disease pathogenesis.
Proceedings of the National Academy of Sciences of the United States of America (2013-06-12)
James C Dodge, Christopher M Treleaven, Jonathan A Fidler, Thomas J Tamsett, Channa Bao, Michelle Searles, Tatyana V Taksir, Kuma Misra, Richard L Sidman, Seng H Cheng, Lamya S Shihabuddin
ABSTRACT
Metabolic dysfunction is an important modulator of disease course in amyotrophic lateral sclerosis (ALS). We report here that a familial mouse model (transgenic mice over-expressing the G93A mutation of the Cu/Zn superoxide dismutase 1 gene) of ALS enters a progressive state of acidosis that is associated with several metabolic (hormonal) alternations that favor lipolysis. Extensive investigation of the major determinants of H(+) concentration (i.e., the strong ion difference and the strong ion gap) suggests that acidosis is also due in part to the presence of an unknown anion. Consistent with a compensatory response to avert pathological acidosis, ALS mice harbor increased accumulation of glycogen in CNS and visceral tissues. The altered glycogen is associated with fluctuations in lysosomal and neutral α-glucosidase activities. Disease-related changes in glycogen, glucose, and α-glucosidase activity are also found in spinal cord tissue samples of autopsied patients with ALS. Collectively, these data provide insights into the pathogenesis of ALS as well as potential targets for drug development.
Sigma-Aldrich
Glycogen from mussel, Mytilus genus, for DNA precipitations
Sigma-Aldrich
Glycogen from mussel, Mytilus genus, ≥85% anhydrous basis (enzymatic)
Sigma-Aldrich
Glycogen from rabbit liver, ≥85% dry basis (enzymatic)
Sigma-Aldrich
Glycogen from bovine liver, ≥85% dry basis (enzymatic)
Sigma-Aldrich
Glycogen from oyster, ≥75% dry basis