Merck
  • Experimental study on calcium carbonate precipitation using electromagnetic field treatment.
Experimental study on calcium carbonate precipitation using electromagnetic field treatment.
Water science and technology : a journal of the International Association on Water Pollution Research (2013-06-22)
Miao Xuefei, Xiong Lan, Chen Jiapeng, Yang Zikang, He Wei
ABSTRACT
The present study investigated the effectiveness of electromagnetic fields in preventing calcium carbonate (CaCO₃) fouling in cooling water. Four different frequencies and two different voltages were adopted to induce electromagnetic fields directly in water with constant water temperature and constant flow velocity. Artificial hard water was used. The solution conductivities decreased by 17-25% from their initial values in the electromagnetic anti-fouling treatment (EAT) cases, depending on different frequencies of electric pulses, whereas the untreated case dropped by 31%. The particle size became small and the crystal structure changed into loose style after EAT. The EAT device independently developed by the State Key Laboratory had been validated as an effective apparatus in preventing CaCO₃ fouling in cooling water.
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
Calcium carbonate, ACS reagent, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Supelco
Calcium carbonate, reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.