Merck
[Membrane-bound proteases of ompT+ and ompT- Vibrio cholerae strains].
Zhurnal mikrobiologii, epidemiologii, i immunobiologii (2013-06-29)
S N Kozlov, V B Nikolaev, E Iu Markov, L Ia Urbanovich, L V Mironova
ABSTRACT
Detection ofproteases in outer membranes (OM) of ompT+ and ompT- Vibrio cholerae strains of O1 and O139 serogroups. Specific sterile preparations of OM were obtained by lysis of live V. cholerae cells by 4.5 M urea solution with subsequent differential centrifugation and treatment by nucleases. Extraction of OM proteins previously treated by sodium sarcosinate was carried out by Triton X-100 in the presence of EDTA. Protease and polypeptide spectra were studied in substrate and SDS electrophoresis. Sensitivity of proteases to inhibitors was determined in diffusion test in agarose gel containing substrate by using soy trypsin inhibitor (STI) and phenylmethylsulfonyl fluoride (PMSF). The presence of ompT was determined in PCR by using specific primers. According to PCR data 13 Vibrio cholerae O1 strains and 3 V. cholerae O139 strains isolated from clinical material as well as 22 V. cholerae O1 strains isolated from environmental objects contained ompT gene. 2 V. cholerae O1 human isolated strains, 9 V. cholerae O1 strains and 2 V. cholerae O139 strains isolated from the environment did not have ompT gene. By using SDS- and enzyme-electrophoresis in polyacrylamide gel quantitative and qualitative differences in composition of polypeptides and proteases of OM ompT+ and ompT- V. cholerae strains that hydrolyze gelatin, casein and protamine sulfate were detected. Inhibition of OM by STI and PMSF resulted in a decrease of their proteolytic activity. In preparations and extracts of ompT+ and ompT- V. cholerae OM up to 3 proteases some of which may be related to ompT-like were detected.
Product Description
Supelco
Ethylenediaminetetraacetic acid disodium salt dihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
Sigma-Aldrich
Gelatin from bovine skin, Type B
Sigma-Aldrich
Triton X-305 solution, 70% in H2O
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Triton X-100 solution, BioUltra, Molecular Biology, ~10% in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Sarcosine, BioXtra
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Sarcosine, crystallized, ≥98.0% (T)
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid calcium disodium salt hydrate
Sigma-Aldrich
Triton X-102
Sigma-Aldrich
Sarcosine, 98%
Octoxinol 10, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
Gelatin, tested according to Ph. Eur.
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)