Merck
  • Catalytic properties of Ancrod, the thrombin-like proteinase from the Malayan pit viper (Agkistrodon rhodostoma) venom.
Catalytic properties of Ancrod, the thrombin-like proteinase from the Malayan pit viper (Agkistrodon rhodostoma) venom.
Biochimica et biophysica acta (1985-07-01)
P Ascenzi, A Bertollini, M Bolognesi, M Guarneri, E Menegatti, G Amiconi
ABSTRACT
Kinetics for the hydrolysis of p-nitrophenyl esters of N-alpha-carbobenzoxy-L-amino acids catalyzed by Ancrod were determined between pH 5 and 10 (I = 0.1 M) at 21 +/- 0.5 degrees C; the results are consistent with the minimum three-step mechanism: (formula: see text) For all substrates examined, the pH profiles of kcat and/or kcat/Km reflect the ionization of two groups with pKa values ranging between 6.9 and 7.2, and 9.3 and 9.6 (probably, the histidine residue involved in the catalytic triad and the N-terminus, respectively); at variance, values of Km are pH-independent. Moreover, the formation of the E X S complexes may be regarded as a pseudo-equilibrium process, and the acylation step (k + 2) is always rate-limiting in catalysis. Among p-nitrophenyl esters examined, ZArgONp shows the most favourable kinetic parameters and may be the substrate of choice for Ancrod, in that it allows the determination of the enzyme concentration as low as 1 X 10(-9) M (approximately equal to 0.1 Ancrod units/ml), at the optimum pH value (approximately equal to 8). The catalytic behaviour of Ancrod is compared to that of serine proteinases acting on cationic and non-cationic substrates; differences in kinetics, which refer to a lower enzyme:substrate affinity, may be related to a higher rigidity, lower hydrophobicity and/or adverse steric hindrance of the S1 subsite of Ancrod.
Sigma-Aldrich
Z-L-Lys-ONp hydrochloride
Pricing and availability is not currently available.