Merck
  • New assay for the O-alkyl cleavage enzyme with alkyl-lysophospholipids as substrates.
New assay for the O-alkyl cleavage enzyme with alkyl-lysophospholipids as substrates.
Cancer research (1985-02-01)
C Unger, H Eibl, H W von Heyden, G A Nagel
ABSTRACT
The O-alkyl cleavage enzyme is important for the metabolism of cytotoxic alkyl-lysophospholipids. We have developed a simple new method for the determination of the enzyme activity which is based on the formation of water-soluble phosphate during the enzyme reaction. This is the first assay which avoids the use of radiolabeled substrates.
Sigma-Aldrich
6,7-Dimethyl-5,6,7,8-tetrahydropterine hydrochloride, ≥95%