Merck
  • Coexistence of different pathways in the metabolism of n-propylbenzene by Pseudomonas sp.
Coexistence of different pathways in the metabolism of n-propylbenzene by Pseudomonas sp.
Applied and environmental microbiology (1979-11-01)
Y Jigami, Y Kawasaki, T Omori, Y Minoda
PMID543699
ABSTRACT
Pseudomonas desmolytica S449B1 and Pseudomonas convexa S107B1 grown on n-propylbenzene oxidized n-propylbenzene to beta-phenylpropionic acid and benzoic acid by initial oxidation of the n-propyl side chain and the following beta-oxidation, respectively. The same strains also oxidized n-propylbenzene to 3-n-propylcatechol by initial oxidation of positions 2 and 3 of the aromatic nucleus. A ring fission product, 2-hydroxy-6-oxononanoic acid, was also isolated from the culture broth. Together with the results of oxygen uptake experiments, the data obtained suggested not only the existence of a reductive step to form 2-hydroxy-6-oxononanoic acid, but also the coexistence of two different pathways in the metabolism of n-propylbenzene by the strains used.
Sigma-Aldrich
Propylbenzene, ≥99.0% (GC)
Supelco
Propylbenzene, analytical standard
Sigma-Aldrich
Propylbenzene, 98%