Merck
  • Two-dimensional NMR spectroscopy of siomycin A. Proton--carbon-13 chemical shift correlation.
Two-dimensional NMR spectroscopy of siomycin A. Proton--carbon-13 chemical shift correlation.
European journal of biochemistry (1982-03-01)
N J Clayden, F Inagaki, R J Williams, G A Morris, K Tori, K Tokura, T Miyazawa
ABSTRACT
A trial application of a recent two-dimensional nuclear magnetic resonance experiment to the polypeptide antibiotic siomycin A is described. Proton--carbon-13 chemical shift correlation measures the proton and carbon-13 chemical shift for each directly bonded CH group in a molecule, in a single experiment. The resultant map of correlated chemical shifts enables the carbon-13 spectrum to be assigned directly from the known proton shifts, and allows individual proton signals to be identified without problems of overlap. The signal-to-noise ratio available from such techniques should enable their application to aqueous protein solutions using currently available high-field spectrometers.