Merck
  • Changes of n-hexane metabolites in urine of rats exposed to various concentrations of n-hexane and to its mixture with toluene or MEK.
Changes of n-hexane metabolites in urine of rats exposed to various concentrations of n-hexane and to its mixture with toluene or MEK.
International archives of occupational and environmental health (1983-01-01)
M Iwata, Y Takeuchi, N Hisanaga, Y Ono
ABSTRACT
It is well known that n-hexane produces peripheral neuropathy, and 2,5-hexanedione, one of the metabolites of n-hexane, is thought to be the main causative agent. Recently, the metabolites of n-hexane in urine have been measured by gas chromatography, and 2,5-hexanedione was proved to be useful for the biological monitoring of n-hexane exposure. In the present experiment, we intended to clarify the change of n-hexane metabolites in the urine of rats exposed to various concentrations of n-hexane and to its mixture with toluene of MEK. In the first experiment, five separate groups of five rats each were exposed to 100, 500, 1000, or 3000 ppm of n-hexane, or fresh air respectively in an exposure chamber for 8 h a day. Urinary samples were gathered during exposure, 16, 24, and 40 h after exposure. Half of each sample was analyzed by gas chromatography after hydrolysis with acid and enzymes, and the other half was analyzed without hydrolysis. 2,5-Dimethylfuran, MBK, 2-hexanol, 2,5-hexanedione, and gamma-valerolactone could be identified as n-hexane metabolites in the urine. The main metabolites were 2-hexanol and 2,5-hexanedione. 2-Hexanol was mostly excreted during exposure, while most of the 2,5-hexanedione was excreted after the end of exposure. The amount of metabolites in the urine correlatively increased with the concentration of n-hexane from 100 to 1000 ppm, but the amount of metabolites scarcely increased when the concentration of n-hexane increased from 1000 to 3000 ppm.(ABSTRACT TRUNCATED AT 250 WORDS)
Sigma-Aldrich
2-Hexanol, 99%
Sigma-Aldrich
γ-Valerolactone, ≥99%, FCC, FG
Sigma-Aldrich
γ-Valerolactone, ReagentPlus®, 99%
Sigma-Aldrich
γ-Valerolactone, natural, ≥95%, FG