Merck
  • Mutagenesis testing of acetyl-tributylcitrate and epoxidized soybean oil.
Mutagenesis testing of acetyl-tributylcitrate and epoxidized soybean oil.
Poultry science (1982-12-01)
J L Heath, M Reilly
ABSTRACT
Mutagenesis testing was conducted on two plasticizers commonly used in plastic wrap manufacture, acetyl-tributylcitrate and epoxidized soybean oil. There is no record of mutagenic testing using a bacterial screening method for these two compounds. The two plasticizers were screened using mutant strains of Salmonella typhimurium. The tests determined that they were not mutagenic.
Sigma-Aldrich
Tributyl O-acetylcitrate, 98%
Supelco
Tributyl 2-acetylcitrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tributyl 2-acetylcitrate, ≥98%, FG