Merck
Metabolic accumulation of hydrocarbons by Acinetobacter species.
Zhonghua Minguo wei sheng wu ji mian yi xue za zhi = Chinese journal of microbiology and immunology (1982-02-01)
S T Chung, W R Finnerty
ABSTRACT
Acinetobacter HO1-N grown on hexadecane as a sole source of carbon and energy was demonstrated to cytoplasmically sequester this hydrocarbon. Cultures grown with hexadecane and heptadecane accumulated intracytoplasmically both hydrocarbons. The accumulation of specific alkanes was non-specific while the oxidation of the pooled hydrocarbon was highly specific. Naphthalene was highly inhibitory to the growth of this microorganism but slowly co-oxidized in the presence of hexadecane.
Product Description
Supelco
Heptadecane, analytical standard
Sigma-Aldrich
Heptadecane, 99%