Merck
  • Inhibition of insulin release by a formamidine pesticide amitraz and its metabolites in a rat beta-cell line: an action mediated by alpha-2 adrenoceptors, a GTP-binding protein and a decrease in cyclic AMP.
Inhibition of insulin release by a formamidine pesticide amitraz and its metabolites in a rat beta-cell line: an action mediated by alpha-2 adrenoceptors, a GTP-binding protein and a decrease in cyclic AMP.
The Journal of pharmacology and experimental therapeutics (1994-12-01)
T H Chen, W H Hsu
ABSTRACT
We tested the hypothesis that the formamidine pesticide amitraz and its metabolites inhibit insulin release from a rat beta-cell line (RINm5F) by decreasing intracellular cyclic AMP levels and examined whether GTP-binding proteins were involved in the process. Amitraz and its active metabolite BTS27271 (0.1-10 microM) inhibited insulin release that was induced by 100 microM of IBMX in a dose-dependent manner. Other metabolites of amitraz tested failed to inhibit insulin release. A potent and specific alpha-2 adrenoceptor agonist, medetomidine (0.1 microM), abolished the IBMX-induced insulin release. Amitraz, BTS27271 and medetomidine also decreased intracellular cyclic AMP concentrations that were elevated by IBMX administration. Moreover, all three drugs inhibited the insulin release stimulated by forskolin (1 microM), an adenylyl cyclase activator. Yohimbine (0.01, 0.1 and 1 microM), an alpha-2 adrenoceptor antagonist, prevented the inhibitory effect of amitraz and BTS27271 on insulin release, whereas prazosin (1 microM), an alpha-1 adrenoceptor antagonist, did not. Yohimbine, but not prazosin, also prevented the effect of amitraz, BTS27271 and medetomidine on IBMX-stimulated accumulation of cyclic AMP. Pretreatment of cells with PTX (0.1 micrograms/ml) for 16 h, antagonized the effects of amitraz and BTS27271 on IBMX-induced increase in insulin release. Thus, one mechanism by which amitraz and its metabolite BTS27271 decrease insulin release is by inhibiting adenylyl cyclase, an action mediated through a PTX-sensitive G-protein.