Merck
  • Improved separation of diastereomeric derivatives of enantiomers by a physical network of linear polyvinylpyrrolidone applied as pseudophase in capillary zone electrophoresis.
Improved separation of diastereomeric derivatives of enantiomers by a physical network of linear polyvinylpyrrolidone applied as pseudophase in capillary zone electrophoresis.
Electrophoresis (1994-06-01)
W Schützner, G Caponecchi, S Fanali, A Rizzi, E Kenndler
ABSTRACT
Diastereomeric analytes were separated using capillary zone electrophoresis with polyvinylpyrrolidone as polymeric additive to the buffer solution. As test substances derivatives of D- and L- alpha-amino and alpha-hydroxy acids formed by reaction with (+)-O,O'-diacetyl- and (+)-O,O'-dibenzoyl-L-tartaric anhydride, respectively, were used. The physical network formed by the linear polymer is supposed to act as a kind of pseudophase. It was found that the network affects the mobility of diastereomeric compounds to a different extent, enhancing the selectivity of the system. In nearly all cases of aromatic acids the diastereomer carrying the D-amino acid was more strongly retained than the L-isomer, as opposed to the situation with aliphatic acids.
Sigma-Aldrich
(+)-O,O′-Diacetyl-L-tartaric anhydride, 97%
Pricing and availability is not currently available.