Merck
  • The use of low-energy collisionally activated dissociation negative-ion tandem mass spectrometry for the characterization of dog and human urinary metabolites of the drug BW 1370U87.
The use of low-energy collisionally activated dissociation negative-ion tandem mass spectrometry for the characterization of dog and human urinary metabolites of the drug BW 1370U87.
Rapid communications in mass spectrometry : RCM (1994-03-01)
L C Taylor, R L Johnson, L St John-Williams, T Johnson, S Y Chang
ABSTRACT
Liquid chromatography (LC) combined with atmospheric pressure chemical ionization mass spectrometry was used to identify phase I and II metabolites of the drug BW 1370U87 in dog and human urine. Additional analysis of individual high-performance liquid chromatograph fractions collected from dog urine by combined gas chromatography/mass spectrometry identified one metabolite which was not detected by LC/MS methods. Using negative-ion LC/MS, the majority of BW 1370U87 metabolites in human urine were identified as glucuronic acid conjugates of phase I oxidative metabolites. The negative-ion fragmentation produced by low-energy collisionally activated decomposition (CAD), studied by tandem mass spectrometry experiments, confirmed that these compounds were drug-related and allowed metabolite structures to be assigned. Product-ion spectra of the metabolites were dominated by the loss of neutral molecules from even-electron deprotonated [M-H]- ions.
Sigma-Aldrich
Butylboronic acid, 97%
Supelco
Butylboronic acid, derivatization grade (GC derivatization), LiChropur, ≥96.0% (T)