Merck
  • Antileukemic activity of phenylalanine methyl ester (PME): a lysosomotropic peptide methyl ester.
Antileukemic activity of phenylalanine methyl ester (PME): a lysosomotropic peptide methyl ester.
Stem cells (Dayton, Ohio) (1994-03-01)
C S Rosenfeld
ABSTRACT
The antileukemic activities of the lysosomotropic compounds, such as phenylalanine methyl ester (PME), have received little attention. In this study, a 3-[4,5-Dimethylthiazol-2-yl]-2,5-diphenyltetrazolium bromide (MTT) assay was used to investigate the antileukemic activity of PME. Leukemic specimens from untreated patients that contained greater than or equal to 75% blasts were used. Leukemic cells were treated with PME at 37 degrees C and 22 degrees C in concentrations ranging from 0.5 to 50 mM. Normal blood mononuclear cells served as controls. At both 37 degrees C and 22 degrees C, the recovery of normal peripheral blood cells was approximately 28% following incubation with 50 mM PME. At 37 degrees C, 50 mM PME caused greater than one log reduction of leukemic cells in 13/16 acute myelogenous leukemia (AML), 7/9 acute lymphocytic leukemia (ALL), and 8/8 in blast crisis of chronic myelogenous leukemia (CML-BC) specimens. PME had less activity at 22 degrees C than at 37 degrees C. PME was compared with 100 micrograms/ml 4-hydroperoxycyclophosphamide (4HC). In contrast to PME, 4HC was associated with a greater than one log reduction of leukemic cells in only 1/13 AML, 1/3 ALL and 0/6 CML-BC specimens. 4HC activity exceeded PME activity in only one case each of ALL and prolymphocytic leukemia (PLL). In a case of CD34+ B cell ALL, synergy of PME and 4HC was demonstrated. These studies indicate 1) PME has antileukemic activity and 2) 4HC has less antileukemic activity than PME.
Sigma-Aldrich
L-Phenylalanine methyl ester hydrochloride, 98%
Sigma-Aldrich
L-Phenylalanine methyl ester hydrochloride
Sigma-Aldrich
D-Phenylalanine methyl ester hydrochloride, 98%