Merck
  • Intrinsic neuromodulation in the Tritonia swim CPG: serotonin mediates both neuromodulation and neurotransmission by the dorsal swim interneurons.
Intrinsic neuromodulation in the Tritonia swim CPG: serotonin mediates both neuromodulation and neurotransmission by the dorsal swim interneurons.
Journal of neurophysiology (1995-12-01)
P S Katz, W N Frost
ABSTRACT
1. Neuromodulation has previously been shown to be intrinsic to the central pattern generator (CPG) circuit that generates the escape swim of the nudibranch mollusk Tritonia diomedea; the dorsal swim interneurons (DSIs) make conventional monosynaptic connections and evoke neuromodulatory effects within the swim motor circuit. The conventional synaptic potentials evoked by a DSI onto cerebral neuron 2 (C2) and onto the dorsal flexion neurons (DFNs) consist of a fast excitatory postsynaptic potential (EPSP) followed by a prolonged slow EPSP. In their neuromodulatory role, the DSIs produce an enhancement of the monosynaptic connections made by C2 onto other CPG circuit interneurons and onto efferent flexion neurons. Previous work showed that the DSIs are immunoreactive for serotonin. Here we provide evidence that both the neurotransmission and the neuromodulation evoked by the DSIs are produced by serotonin, and that these effects may be pharmacologically separable. 2. Previously it was shown that bath-applied serotonin both mimics and occludes the modulation of the C2 synapses by the DSIs. Here we find that pressure-applied puffs of serotonin mimic both the fast and slow EPSPs evoked by a DSI onto a DFN, whereas high concentrations of bath-applied serotonin occlude both of these synaptic components. 3. Consistent with the hypothesis that serotonin mediates the actions of the DSIs, the serotonin reuptake inhibitor imipramine prolongs the duration of the fast DSI-DFN EPSP, increases the amplitude of the slow DSI-DFN EPSP, and increases both the amplitude and duration of the modulation of the C2-DFN synapse by the DSIs. 4. Two serotonergic antagonists were found that block the actions of the DSIs. Gramine blocks the fast DSI-DFN EPSP, and has far less of an effect on the slow EPSP and the modulation. Gramine also diminishes the depolarization evoked by pressure-applied serotonin, showing that it is a serotonin antagonist in this system. In contrast, methysergide greatly reduces both the slow EPSP and the modulation evoked by the DSIs, but has mixed effects on the fast EPSP. Methysergide also blocks the ability of exogenous serotonin to enhance the C2-DFN EPSP, demonstrating that it antagonizes the serotonin receptors responsible for this modulation. 5. Taken together with previous work, these results indicate that serotonin is likely to be responsible for all three actions of the DSIs that were examined: the fast and slow DSI-DFN EPSPs and the neuromodulation of the C2-DFN synapse. These results also indicate that the conventional and neuromodulatory effects of the DSIs may be pharmacologically separable. In future work it may be possible to determine the functional role of each in the swim circuit.
Sigma-Aldrich
Gramine, 97.5%