Merck
[The corticostriatal mechanisms of behavior].
Fiziologicheskii zhurnal imeni I.M. Sechenova (1995-12-01)
N F Suvorov, V T Shuvaev, N L Voĭlokova, O G Chivileva, V I Shefer
ABSTRACT
Experiments in dogs, cats and monkeys revealed that, along with the diffuse principle of afferent terminal arrangement within the striatum, there exist some features of terminal organisation by the anterior-posterior and medio-lateral gradients. The data obtained suggest that the prefrontal cortex and the caudate nucleus head maintain programming of intentions and the evaluation of performed actions.
Sigma-Aldrich
L-Glutamic acid diethyl ester hydrochloride, 97%